missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ UltraGrip™ Boot Covers with Elastic Ankle

Product Code. 12765935
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
Universal
X-Large
Thread Color:
Black
White
Yellow
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Thread Color unitSize
12765935 Universal White Each
12755935 Medium Yellow -
12775935 X-Large Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12765935 Supplier AlphaProTech Supplier No. BTA1812B

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-top boot covers feature soft, strong, and flexible Microbreathe™ material with a high vapor transmission rate for all day comfort.

  • Microbreathe upper material provides barrier protection
  • Static dissapative
  • High vapor transmission rate

Specifications

Size Universal
Material Polypropylene
Latex Free Yes
Color White
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Anti-skid
Seam Type Serged
Thread Color White
Certifications/Compliance ASTM D751
Packaging Quantity 100 Pairs/Cs.
Dimensions (L x W x H) 14 x 4.5 x 17.5 in. (35.56 x 11.43 x 44.45cm)
Length (English) 14 in.
Length (Metric) 35.56 cm
Height (English) 17.5 in.
Height (Metric) 44.45 cm
Product Type Controlled Environments Boot Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.