missing translation for 'onlineSavingsMsg'
Learn More

(AlphaR)-2-Chloro-Alpha-[[(4-methylphenyl)sulfonyl]oxy]benzeneacetic Acid Methyl Ester, TRC

Product Code. 30353875
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30353875 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353875 Supplier Toronto Research Chemicals Supplier No. TRCC3750752.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (alphaR)-2-Chloro-alpha-(((4-methylphenyl)sulfonyl)oxy)benzeneacetic Acid Methyl Ester
CAS 223123-53-3
Molecular Formula C16H15ClO5S
Quantity 2.5 g
InChI Formula InChI=1S/C16H15ClO5S/c1-11-7-9-12(10-8-11)23(19,20)22-15(16(18)21-2)13-5-3-4-6-14(13)17/h3-10,15H,1-2H3/t15-/m1/s1
SMILES COC(=O)[C@H](OS(=O)(=O)c1ccc(C)cc1)c2ccccc2Cl
IUPAC Name methyl (2R)-2-(2-chlorophenyl)-2-(4-methylphenyl)sulfonyloxyacetate
Molecular Weight (g/mol) 354.81
Formula Weight 354.0329
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.