missing translation for 'onlineSavingsMsg'
Learn More

Skytec™ Alphashield 1000-S1BH Disposable Coverall

Product Code. 31702866
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31702866 Small Pack of 50
31702861 Medium Pack of 50
31702854 Large Pack of 50
31702868 X-Large Pack of 50
31702872 2X-Large Pack of 50
31702850 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31702866 Supplier Skytec™ Supplier No. PDS1B/HS1000/WH1
  • ONLINE EXCLUSIVE PRICE
    656.13 €
    420.37 € / Pack of 50
    Save 235.76 €
    35% Off
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lightweight disposable coverall designed to protect workers and prevent contamination in sensitive environments.

  • Three-piece elasticated hood construction provides added comfort, durability, and fit
  • Elasticated ankles and wrists
  • Storm flap covering zip offers reassuring protection
  • Quick and easy donning/doffing with longer 2-way zip
  • Elasticated back waist for enhanced comfort

Specifications

Product Type Chemical-resistant Coveralls
Size Small
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals
Certifications/Compliance CE III or - UKCA or - EN 1073-2:2002 CLASS 1 or - EN 1149-5:2018 or - EN 13034:2005 + A1:2010 Type 6 or - EN 13982-1:2004 + A1:2010 Type 5
For Use With (Application) Farm Maintenance, Biohazards, Chemical Handling and Containment, Cleanroom, Electrical Engineering and Maintenance
Weight (Metric) 167 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.