Learn More
Skytec™ Alphashield 2000+- S2BH+ Disposable Coverall
Product Code. 31702870
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31702870 Large Pack of 50
31702851 Small Pack of 50
31702863 Medium Pack of 50
31702862 X-Large Pack of 50
31702856 2X-Large Pack of 50
31702873 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31702870 Supplier Skytec™ Supplier No. PDS2B/HS200+/WH3
  • ONLINE EXCLUSIVE PRICE
    638.44 €
    409.05 € / Pack of 50
    Save 229.39 €
    35% Off
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Limited life disposable coverall made from lightweight material with a breathable film, designed to protect workers and prevent contamination in sensitive environments.
  • Spun bonded polypropylene with outer breathable film with chemical liquid (EN368 and blood (ASTM F1670) penetration resistance for premium protection
  • Strong, durable, and comfortable
  • Improved fit with two-piece crotch pattern
  • Quick and easy donning/doffing in an emergency with longer 2-way zip
  • Fully elasticated hood, ankles, and wrists
Specifications
Product Type Chemical-resistant Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals
Certifications/Compliance CE III or - UKCA or - EN 1073-2:2002 CLASS 1 or - EN 1149-5:2018 or - EN 13034:2005 + A1:2010 Type 6 or - EN 13982-1:2004 + A1:2010 Type 5 or - EN 14126:2003 + AC:2004 Type 5B,Type 6B
For Use With (Application) Farm Maintenance, Biohazards, Chemical Handling and Containment, Cleanroom, Electrical Engineering and Maintenance, Painting and Finishing
Weight (Metric) 450 g