Learn More
Skytec™ Alphashield 2000+- S2BH+ Disposable Coverall
Product Code. 31702873 Shop All Skytec Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31702873 3X-Large Pack of 50
31702851 Small Pack of 50
31702863 Medium Pack of 50
31702870 Large Pack of 50
31702862 X-Large Pack of 50
31702856 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31702873 Supplier Skytec™ Supplier No. PDS2B/HS200+/WH6
  • ONLINE EXCLUSIVE PRICE
    638.44 €
    409.05 € / Pack of 50
    Save 229.39 €
    35% Off
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Limited life disposable coverall made from lightweight material with a breathable film, designed to protect workers and prevent contamination in sensitive environments.
  • Spun bonded polypropylene with outer breathable film with chemical liquid (EN368 and blood (ASTM F1670) penetration resistance for premium protection
  • Strong, durable, and comfortable
  • Improved fit with two-piece crotch pattern
  • Quick and easy donning/doffing in an emergency with longer 2-way zip
  • Fully elasticated hood, ankles, and wrists
Specifications
Product Type Chemical-resistant Coveralls
Size 3X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals
Certifications/Compliance CE III or - UKCA or - EN 1073-2:2002 CLASS 1 or - EN 1149-5:2018 or - EN 13034:2005 + A1:2010 Type 6 or - EN 13982-1:2004 + A1:2010 Type 5 or - EN 14126:2003 + AC:2004 Type 5B,Type 6B
For Use With (Application) Farm Maintenance, Biohazards, Chemical Handling and Containment, Cleanroom, Electrical Engineering and Maintenance, Painting and Finishing
Weight (Metric) 600 g