Learn More
Skytec™ Alphashield 2000CP-S2BH-CP Disposable Coverall
Product Code. 31702871 Shop All Skytec Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31702871 2X-Large Pack of 50
31702853 Small Pack of 50
31702859 Medium Pack of 50
31702855 Large Pack of 50
31702852 X-Large Pack of 50
31702860 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31702871 Supplier Skytec™ Supplier No. PDS2B/HS200C/WH5
  • ONLINE EXCLUSIVE PRICE
    722.06 €
    462.62 € / Pack of 50
    Save 259.44 €
    35% Off
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Limited life disposable coverall made from lightweight material with a breathable film, designed to protect workers and prevent contamination in sensitive environments.
  • Lightweight spun-bonded polypropylene with outer breathable film
  • Back comfort panel and gusset area
  • Taped sealing on storm flap for extra protection
  • Thumb loop ensures arm protection remains in place when the arm is raised
  • Three-piece elasticated hood construction provides added comfort, durability, and fit
  • Elasticated ankles and wrists
Specifications
Product Type Chemical-resistant Coveralls
Size 2X-Large
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals
Certifications/Compliance CE III or - UKCA or - EN 1073-2:2002 CLASS 1 or - EN 1149-5:2018 or - EN 13034:2005 + A1:2010 Type 6 or - EN 13982-1:2004 + A1:2010 Type 5
For Use With (Application) Farm Maintenance, Biohazards, Chemical Handling and Containment, Cleanroom, Electrical Engineering and Maintenance, Painting and Finishing
Weight (Metric) 550 g