Learn More
Skytec™ AlphaShield - S2200 Disposable Coverall
Product Code. 31702865 Shop All Skytec Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31702865 X-Large Pack of 50
31702869 Small Pack of 50
31702874 Medium Pack of 50
31702857 Large Pack of 50
31702867 2X-Large Pack of 50
31702864 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31702865 Supplier Skytec™ Supplier No. PDS2B/HS2200/WH4
  • ONLINE EXCLUSIVE PRICE
    564.46 €
    361.65 € / Pack of 50
    Save 202.81 €
    35% Off
View all promotions
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Limited life disposable coverall with bound seam, designed to protect workers and prevent contamination in sensitive environments.
  • Lightweight microporous polyethylene (PE) laminate
  • Bound seam for additional protection and strength
  • 2-way front zipper with re-sealable storm flap
  • Thumb loop ensures arm protection remains in place when arm is raised
  • Elasticated waist to enhance fit
  • Innovative three-piece elasticated hood construction provides added comfort, durability and fit
  • Elasticated ankles and wrists
Specifications
Product Type Chemical-resistant Coveralls
Size X-Large
Gender Unisex
Material Polyethylene, Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals
Certifications/Compliance CE III or - EN 1073-2:2002 CLASS 1 or - EN 1149-5:2018 or - EN 13034:2005 + A1:2010 Type 6 or - EN 13982-1:2004 + A1:2010 Type 5 or - EN 14126:2003 + AC:2004 Type 3B,Type 4B,Type 5B,Type 6B
For Use With (Application) Farm Maintenance, Biohazards, Chemical Handling and Containment, Cleanroom, Electrical Engineering and Maintenance, Painting and Finishing
Weight (Metric) 315 g