Learn More
Skytec™ AlphaShield - S2200 Disposable Coverall
Product Code. 31702869
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31702869 Small Pack of 50
31702874 Medium Pack of 50
31702857 Large Pack of 50
31702865 X-Large Pack of 50
31702867 2X-Large Pack of 50
31702864 3X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31702869 Supplier Skytec™ Supplier No. PDS2B/HS2200/WH1
  • ONLINE EXCLUSIVE PRICE
    564.46 €
    361.65 € / Pack of 50
    Save 202.81 €
    35% Off
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Limited life disposable coverall with bound seam, designed to protect workers and prevent contamination in sensitive environments.
  • Lightweight microporous polyethylene (PE) laminate
  • Bound seam for additional protection and strength
  • 2-way front zipper with re-sealable storm flap
  • Thumb loop ensures arm protection remains in place when arm is raised
  • Elasticated waist to enhance fit
  • Innovative three-piece elasticated hood construction provides added comfort, durability and fit
  • Elasticated ankles and wrists
Specifications
Product Type Chemical-resistant Coveralls
Size Small
Gender Unisex
Material Spunbond Polypropylene
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Anti-static, Particle, Liquid, Chemicals
Certifications/Compliance CE III or - EN 1073-2:2002 CLASS 1 or - EN 1149-5:2018 or - EN 13034:2005 + A1:2010 Type 6 or - EN 13982-1:2004 + A1:2010 Type 5 or - EN 14126:2003 + AC:2004 Type 3B,Type 4B,Type 5B,Type 6B
For Use With (Application) Farm Maintenance, Biohazards, Chemical Handling and Containment, Cleanroom, Electrical Engineering and Maintenance, Painting and Finishing
Weight (Metric) 274 g