missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 02-100 Laminated Safety Gloves
Description
- Specialized protection: AlphaTec™ 02-100 laminated safety gloves have a multi-layered design, protecting against a broad range of chemicals, meeting EN Type A chemical resistance standards and ensuring greater workplace safety
- Advanced defenses: Beyond chemical protection certification, they possess EN 374 viral protection capabilities
- Guaranteed quality: AlphaTec™ 02-100 industrial gloves are also rigorously air-pressure tested and 100% inspected, ensuring exceptional quality, for truly reliable personal protection
Specifications
Specifications
| Applications | Industrial |
| Color | White |
| Latex Free | Yes |
| Material | LLDPE Laminated Film |
| Product Type | Chemical-resistant Gloves |
| Size | 10 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?