Learn More
Ansell Alphatec™ 1500 Plus Lab Coat Bound - Model 204
Product Code. 15952931 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 60
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15952931 Large Pack of 60
15942931 Medium Pack of 60
15962931 X-Large Pack of 60
15972931 2X-Large Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15952931 Supplier Ansell Supplier No. WH15B0020404
Estimated Shipment: 27-04-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Alphatec™ 1500 Plus Lab Coat delivers comfortable protection in general industrial environments.
  • Optimized breathability: AlphaTec™ 1500 PLUS Lab Coat Bound - Model 204 delivers comfortable protection in general industrial environments, thanks to its air and water vapor permeable as well as anti-static, SMS material
  • Reduced contamination risks: As this protective outer garment is silicone-free, it minimizes contamination risks, making it ideal for the life sciences and metal fabrication industries
  • Improved ergonomics: In addition, this white protective over garment offers an optimized fit, which improves wearer comfort and workplace safety
DESIGN FEATURES:
  • Medium length labcoat with collar, studded front fastening, left breast and right thigh pockets.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Anti-static Lab Coat
Size Large
Gender Unisex
Material SMS
Color White
Closure Type Buttons
Cuff Style Elastic
Disposable Reusable
Sterility Non-sterile
Resistance Chemicals
Style Below Hip-length
No. of Pockets 2
Latex Free Yes
Weight (English) 48 gsm