missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 1500 STANDARD Stitched Coveralls, Model 138, White

Product Code. 18050140 Shop All Ansell Products
Change view
Click to view available options
Size:
4X-Large
6X-Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18050140 6X-Large Each
18070140 4X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18050140 Supplier Ansell Supplier No. WH15S0013810
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Provides comfortable protection from hazardous particulates in general industrial environments.

SUIT FEATURES

  • 3 piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size 6X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class Category III risks I Type 5/6
Resistance Chemicals, Particle
Collar Type 3-piece elasticated hood
Certifications/Compliance (Europe) CE-nummer|EN ISO 13688:2013|EN ISO 13982-1:2004 + A1:2010|EN ISO 13982-1:2004 + A1:2010|EN ISO 13982-1:2004 + A1:2010|EN 13034:2005 + A1:2009|EN 1073-2:2002
Latex Free Yes
For Use With (Application) Inspection (picking, checking, and marking), Asbestos Related Work, Handling of powders, General maintenance
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.