Learn More
Ansell AlphaTec™ 1500 STANDARD Stitched, Model 138, Navy
Product Code. 18030120 Shop All Ansell Products
Change view
Click to view available options
Size:
4X-Large
5X-Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18030120 5X-Large Each
18050120 4X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18030120 Supplier Ansell Supplier No. NV15S0013809
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Provides comfortable protection from hazardous particulates in general industrial environments.
SUIT FEATURES:
  • 3 piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size 5X-Large
Gender Unisex
Material SMS Fabric
Color Navy
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class CE CATEGORY III (Type 5, Type 6)
Resistance Chemicals, Particle
Collar Type Elasticated hood
Certifications/Compliance (Europe) CE Number|EN ISO 13688:2013|EN ISO 13982-1:2004 + A1:2010|EN ISO 13982-1:2004 + A1:2010|EN ISO 13982-1:2004 + A1:2010|EN 13034:2005 + A1:2009|EN 1073-2:2002
Latex Free Yes