missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Apron
Description
- Silicone Free
- Low Linting
Specifications
Specifications
| For Use With (Application) | Clean Room Cleaning and Preparing, Production Line Support and Maintenance, Maintenance of Plant and Machines, Veterinary Services |
| Style | W20B214 |
| Product Type | Controlled Environments Gown |
| Material | Non-woven Microporous Polyethylene Laminate |
| Color | White |
| Closure Type | Tear-back |
| Disposable | Limited-use |
| Sterility | Non-sterile |
| Seam Type | Bound |
| Sleeve Style | Long Sleeve |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?