Learn More
Ansell AlphaTec™ 2000 STANDARD Apron
Product Code. 16130551 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 60
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16130551 Small Pack of 60
16110561 X-Large Pack of 60
16120561 2X-Large Pack of 60
16130561 3X-Large Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16130551 Supplier Ansell Supplier No. WH20B0021402
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 12-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Apron with protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards.
  • Silicone Free
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifications
Type Chemical-resistant Apron
Size Small
Material Non-woven Microporous Polyethylene Laminate
Color White
Disposable Limited-use
Closure Type Tear-back
Sterility Non-sterile
Certifications/Compliance EN ISO 13688:2013, Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003
Weight (Metric) 63 gsm
Style W20B214
Seam Type Bound
Sleeve Style Long Sleeve