Learn More
Ansell AlphaTec™ 2000 STANDARD Apron
Product Code. 16604423
Change view
Click to view available options
Size:
Small
Universal
X-Large
Style:
W20B214
W20S213
Unit Size:
Pack of 200
Pack of 60
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Style unitSize
16604423 Universal W20S213 Pack of 200
16130551 Small W20B214 Pack of 60
16110561 X-Large W20B214 Pack of 60
16120561 - W20B214 Pack of 60
16130561 - W20B214 Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16604423 Supplier Ansell Supplier No. WH20S0021300
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Apron with protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards.
  • Silicone Free
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
For Use With (Application) Clean Room Cleaning and Preparing, Production Line Support and Maintenance, Maintenance of Plant and Machines, Veterinary Services
Style W20S213
Product Type Controlled Environments Gown
Material Non-woven Microporous Polyethylene Laminate
Color White
Closure Type Ties
Disposable Single-use
Sterility Non-sterile
Seam Type Sewn