missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 2000 STANDARD Apron with Sleeves Bound - Model 214

Product Code. 16100561 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
Unit Size:
Pack of 60
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16100561 Large Pack of 60
16190551 Medium Pack of 60
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16100561 Supplier Ansell Supplier No. WH20B0021404
  • ONLINE EXCLUSIVE PRICE
    761.30 €
    498.28 € / Pack of 60
    Save 263.02 €
    34% Off
View all promotions
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AlphaTec™ Apron with Sleeves Bound - Superior breathable microporous laminate technology

  • Proven protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards
  • Silicone Free
  • Low Linting
TRUSTED_SUSTAINABILITY

Specifications

Type Apron
Size Large
Material Microporous Laminate
Color White
Disposable Limited-use
Closure Type Hook and Loop (Rear)
Sterility Non-sterile
For Use With (Application) Critical and Controlled Environments
Style AlphaTec 2000-214
Seam Type Bound
Certifications/Compliance (Europe) EN 14126
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.