missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Bound
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
8X-Large
Large
Medium
One Size
Small
X-Large
X-Small
Style:
W20B102
W20B103
W20B111
W20B113
W20B122
W20B162
Color:
White
White, Green
missing translation for 'unitSize'
Pack of 30
Pack of 35
Pack of 40
Beschreibung
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications
Spezifikation
Spezifikation
| Size | X-Large |
| Material | Microporous Polyethylene Laminate (Non-woven) |
| Color | White |
| Seam Type | Bound |
| Certifications/Compliance (Europe) | EN 374:2003, EN ISO 13982-1:2004 + A1:2010,EN 13034:2005 + A1:2009,EN 1073-2:2002,EN 1149-5:2018,EN 14126:2003,EN ISO 13688:2013,DIN 32781:2010 |
| For Use With (Application) | Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support and Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Crime Scene Investigation (CSI), Veterinary Services |
| Weight (Metric) | 63 gsm |
| Style | W20B113 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur