Learn More
Ansell AlphaTec™ 2000 STANDARD Bound
Product Code. p-7228376
Click to view available options
Size:
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
8X-Large
Large
Medium
One Size
Small
X-Large
X-Small
Style:
W20B102
W20B103
W20B111
W20B113
W20B122
W20B162
Color:
White
White, Green
Unit Size:
Pack of 30
Pack of 35
Pack of 40
Product Code. 16864750
Brand: Ansell WH20B0010202
Only null left
Add to basket
Add to basket
Chemical protective suits that have a proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards.
  • Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone free, critical in spray painting applications
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Seam Type Bound
Certifications/Compliance (Europe) EN 374:2003, EN ISO 13982-1:2004 + A1:2010,EN 13034:2005 + A1:2009,EN 1073-2:2002,EN 1149-5:2018,EN 14126:2003,EN ISO 13688:2013,DIN 32781:2010
For Use With (Application) Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support and Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Crime Scene Investigation (CSI), Veterinary Services
Weight (Metric) 63 gsm
Style W20B102