missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 Ts PLUS
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
W20T103
W20T111
W20T122
Unit Size:
Pack of 30
Pack of 40
Pack of 45
Description
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications
Specifications
Specifications
| Size | Small |
| Material | Microporous Polyethylene Laminate (Non-woven) |
| Color | White |
| Seam Type | Stitched, Taped |
| Certifications/Compliance (Europe) | EN 374:2003, , EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2018, EN 14126:2003, EN ISO 13688:2013, DIN 32781:2010 |
| Includes | Hood and Boots |
| For Use With (Application) | Product Inspection, Loading and Hauling, Fibre Glass/Resin Applications/Ceramic Fibres, Body Painting Including Surface Inspection, Cleaning of Plant and Machines, Maintaining Lab Furniture and Equipment |
| Weight (Metric) | 63 gsm |
| Style | W20T122 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction