Learn More
Ansell AlphaTec™ 2300 STANDARD Bound
Product Code. 16835070
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
Y23B103
Y23B111
Unit Size:
Pack of 40
Product Code. 16835070
Brand: Ansell YY23B0011108
Only null left
Add to basket
Add to basket
Chemical protective suits with a good protective barrier to numerous inorganic liquid chemicals including acids and bases.
  • Lightweight, yet relatively strong and durable
  • Designed to protect, typical coverall features include respirator fit hood and a zip flap with self-adhesive tape closure
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Size 4X-Large
Material Polyethylene on Bicomponent Polypropylene, Polyethylene Non-Woven
Color Yellow
Seam Type Bound
Certifications/Compliance (Europe) EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003
For Use With (Application) Environmental Clean-up and Remediation, Assembly and Disassembly of Large Pipes, Transferring Liquids and Solids, Assembling and Mounting Components
Weight (Metric) 65 gsm
Style Y23B111