missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2300 STANDARD Bound
Description
- Lightweight, yet relatively strong and durable
- Designed to protect, typical coverall features include respirator fit hood and a zip flap with self-adhesive tape closure
- Anti-static
Specifications
Specifications
| Size | Small |
| Material | Polyethylene on Bicomponent Polypropylene, Polyethylene Non-Woven |
| Color | Yellow |
| Seam Type | Bound |
| Certifications/Compliance (Europe) | EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003 |
| For Use With (Application) | Environmental Clean-up and Remediation, Assembly and Disassembly of Large Pipes, Transferring Liquids and Solids, Assembling and Mounting Components |
| Weight (Metric) | 65 gsm |
| Style | Y23B103 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?