Learn More
Ansell AlphaTec™ 2500 PLUS
Product Code. 16834830
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Pack of 30
Product Code. 16834830
Brand: Ansell WH25T0012203
Only null left
Add to basket
Add to basket
Chemical protective suits with durable and breathable microporous laminate technology and Type 3/4/5 protection.
  • Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
  • Ultra low linting to help reduce the risk of cross contamination in critical areas
  • Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
  • Type 3-B liquid tight certified
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Material Microporous Non-woven Polypropylene Laminate
Color White
Seam Type Stitched, Taped
Certifications/Compliance (Europe) EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003
For Use With (Application) Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments
Weight (Metric) 65 gsm
Style W25T122