missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2500 STANDARD
Description
- Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
- Ultra low linting to help reduce the risk of cross contamination in critical areas
- Achieves the highest classifications for protection from biological agents in accordance with EN 14126 and ASTM F 1671 for penetration of blood, body fluids and blood-borne pathogens
- Anti-static
Specifications
Specifications
| Color | White |
| For Use With (Application) | Transferring Liquids and Solids, Unexpected Leakages Spills or Other Releases, Plant Disinfection and Sanitization, Tank Cleaning, Emergency Response, Spill or Leakage Cleanup, Maintenance, Contaminated Environments |
| Hood Type | Attached |
| Material | Microporous Non-woven Polypropylene Laminate |
| Size | Large |
| Product Type | Ultrasonically Welded Coverall |
| Seam Type | Welded |
| Style | W25W122 |
| Weight (Metric) | 65 gsm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?