Learn More
Ansell Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded - Model 243-G01
Product Code. 16844940
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 25
Pack of 30
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16844940 Large Pack of 30
16824940 Small Pack of 30
16834940 Medium Pack of 30
16854940 X-Large Pack of 30
16864940 2X-Large Pack of 25
16874940 3X-Large Pack of 25
16884940 4X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 16844940 Supplier Ansell Supplier No. YE30T0024304G01
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded has rear velcro fastening at the neck.
DESIGN FEATURES:
  • Apron with sleeves with rear velcro fastening at the neck and attached AlphaTec™ 37-676
RECOMMENDED FOR:
  • Unexpected leakages spills or other releases
  • Loading and unloading trucks and vehicles
  • Loading trucks and vehicles
  • Product inspection
  • Cleaning of plant and machines
TRUSTED_SUSTAINABILITY
Specifications
Type Chemical-resistant Apron
Size Large
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Reusable
Closure Type Hook and Loop
Sterility Non-sterile
Weight (Metric) 80 gsm
Seam Type Welded
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126)