Learn More
Ansell™ AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and Taped
Product Code. 31114776
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
31114776 Medium
31114775 Small
31114777 Large
31114778 X-Large
31114779 2X-Large
31114780 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 31114776 Supplier Ansell™ Supplier No. YE30T0021403
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Multi-layer chemical barrier, protects against numerous chemicals and enhances workplace safety
  • The AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and taped - Model 214 chemical protective suit is made with a multi-layer barrier fabric, which offers wearers excellent protection against several chemicals, including inorganic and biological hazards
  • These highly visible, bright yellow chemical protection coveralls improve worker safety in dimly lit environments
  • Featuring welded and taped seams, this full chemical protection suit offers a strong barrier against harmful liquid and particles
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Medium
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Closure Type Hook and Loop (Rear)
Sterility Non-sterile
Latex Free Yes
No. of Pockets None
Seam Type Welded and Taped
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126 / ASTM F1671 - For US only)