Learn More
Ansell™ AlphaTec™ 3000 AVANT2 AIRline Ultrasonically Welded and Taped
Product Code. 16894990
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16894990 Medium Pack of 4
16815000 Large Pack of 4
16835000 X-Large Pack of 4
16855000 2X-Large Pack of 4
16875000 3X-Large Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16894990 Supplier Ansell™ Supplier No. YE30T00075603
Estimated Shipment: 03-06-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AlphaTec™ 3000 AVANT2 AIRline Ultrasonically welded and taped is an encapsulated suit designed for use in combination with the permanently attached AlphaTec™ and AVANT STS V.1 airline regulator.
Encapsulated suit designed to be used with the permanently attached AlphaTec™ and AVANT STS V.1 airline regulator.
  • Includes internal air-channel ventilation system 
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Color Yellow
Hood Type Attached, Elastic Face
Material Multi-layer Barrier Laminate Fabric
Size Medium
Ankle Style Attached Socks
Closure Type Zipper
Cuff Style Double
Disposable Reusable
For Use With (Equipment) Opening and draining pumps valves or lines Filling, blending and charging of raw materials Contaminated Environments Chemical Handling Transferring liquids and solids Loading and unloading of proces equipment Raw material sample collection Concentration mixing and palletisation Hazmat response Assembly and disassembly of large pipes
Gender Unisex
Neck Style Hooded
No. of Pockets None
Product Type Chemical Protective Clothing
Protection Class CE CATEGORY III
Resistance Anti-static
Seam Type Welded
Sterility Non-sterile
Waist Style Non-elastic