missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped - Model 757

Product Code. 17111095
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 4
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17111095 Small Pack of 4
17121095 Small Pack of 4
17141095 Medium Pack of 4
17151095 Medium Pack of 4
17171095 Large Pack of 4
17181095 Large Pack of 4
17101105 X-Large Pack of 4
17111105 X-Large Pack of 4
17131105 2X-Large Pack of 4
17141105 2X-Large Pack of 4
17161105 3X-Large Pack of 4
17171105 3X-Large Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 17111095 Supplier Ansell Supplier No. YE30TA0075702
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped Chemical Protective Suit - Model 757 with HEPA filter offers secondary protection from airline contamination and a SMC silencer maintains the noise within a suit below 70 dB (even at maximum airflow).

  • Specialized protection: The AlphaTec™ 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 757 full body protection suit has been tested against 200 chemicals.
  • Genuine versatility: This chemical protective suit's HEPA filter offers secondary protection from airline contamination and a SMC silencer maintains the noise within a suit below 70 dB (even at maximum airflow)
  • Considered aesthetics: In addition, this protective suit has attached socks with an elasticated boot overflap and is designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
  • Reduced risks: The emergency ripcord feature of this full protective suit permits quick doffing of the suit in cases of emergency or undue distress to the wearer
DESIGN FEATURES:
  • Fully encapsulated suit designed for use in combination with the permanently attached AlphaTec™ AVANT STS V.1 airline regulator.
  • Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop, G01 Ansell Solvex (Nitrile) gloves, G02 Ansell Barrier gloves or G05 AlphaTec™ 29-500 gloves.
  • Attached socks with elasticated boot over-flap and emergency rip cord.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Color Yellow
For Use With (Application) Opening and draining pumps valves or lines, Filling, blending and charging of raw materials, Contaminated Environments, Chemical Handling, Transferring liquids and solids, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Loading and unloading of proces equipment
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size Small
Ankle Style Attached Socks, Elasticated Boot Over Flap
Disposable Reusable
For Use With (Equipment) Air-Fed/Supplied Systems
Gender Unisex
Neck Style Hood
Product Type Encapsulated Chemical Protective Suit
Protection Class CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Seam Type Welded and Taped
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.