Learn More
Ansell™ AlphaTec™ 3000 Overshoes Ultrasonically Welded and Taped
Product Code. 31114787 Shop All Ansell Products
Change view
Click to view available options
Closure Type:
Elastic Opening
Product Code. Closure Type
31114787 Elastic Opening
1 options
Product Code. 31114787 Supplier Ansell™ Supplier No. YE30T0040000
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Multi-layer barrier fabric for chemical-resistant work footwear with flexible and long-lasting personal protection
  • Composed of protective multi-layer barrier fabric, AlphaTec™ 3000 Overshoes Ultrasonically Welded Taped Model 400 affords superb resistance against inorganic chemicals and biological hazards
  • Bright yellow in color, these overshoes not only offer chemical resistance but also high visibility in dark environments to boost worker safety
  • In addition, they are among the most comfortable, lightweight and robust protective footwear on the market
TRUSTED_SUSTAINABILITY
Specifications
Size One Size
Material Multi-layer Barrier Laminate Fabric
Latex Free Yes
Color Yellow
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Opening
Sole Type Standard
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126
Resistance Chemical-resistant, Bloodborne Pathogens, Light Chemical Splash, Airborne Solid Particulates
Product Type Shoe Covers