missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 3000 Ultrasonically Welded Model 122
Click to view available options
Size:
4X-Large
5X-Large
Unit Size:
Each
Description
- Specialized protection: Made with durable multi-layer nonwoven laminate, the AlphaTec™ 3000 Ultrasonically Welded, Model 122 protects against various inorganic chemicals and biological hazards, while ensuring lightweight comfort
- Enhanced features: This chemical-protective suit features dual-zip systems, double cuffs and attached barrier gloves, enhancing its protective capabilities, as well as a two-piece hood and an elasticated waist, sleeves and over-cuffs
- Improved visibility: Thanks to its high-visibility yellow exterior, this protective suit ensures greater wearer and workplace safety
- Assured antistatic capabilities: This full-body protection suit is also tested against EN 1149-5 antistatic standards
Specifications
Specifications
| Product Type | Chemical Protective Clothing |
| Size | 4X-Large |
| Gender | Unisex |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Disposable | Reusable |
| Waist Style | Elastic |
| Ankle Style | Attached Socks, Boot Overflaps |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction