missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CP-673451, 95%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | CP-673451 |
| CAS | 343787-29-1 |
| Synonym | 1-[2-[5-(2-Methoxyethoxy)-1 H-benzimidazol-1-yl]-8-quinolinyl]-4-piperidinamine |
| Quantity | 100 mg |
| Molecular Formula | C24 H27 N5 O2 |
| Formula Weight | 417.51 |
| Physical Form | Solid |
| Product Type | Reagent |
| Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction