Learn More
Ansell AlphaTec™ 4000
Product Code. 17170675
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
G40T103
G40T111
G40T121
G40T125B
G40T125D
G40T132
G40T151
G40T151B
G40T162
Unit Size:
Pack of 10
Pack of 12
Pack of 8
Product Code. 17170675
Brand: Ansell GR40T0016204
Only null left
Add to basket
Add to basket
A chemical protective suit that is engineered to provide an exceptional barrier against a wide range of organic and inorganic chemicals and biological agents.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Anti-static
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Seam Type Welded, Taped
Certifications/Compliance (Europe) EN 374:2003, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 1073-2:2002, EN 1149-5:2008, EN 14126:2003
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes
Weight (Metric) 103 gsm
Style G40T162