missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ GW3965 hydrochloride, 95%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | GW3965 hydrochloride |
| CAS | 405911-17-3 |
| Synonym | 3-[3-[N-(2-Chloro-3-trifluoromethylbenzyl)-(2,2-diphenylethyl)amino]propyloxy]phenylacetic acid hydrochloride |
| Quantity | 100 mg |
| Molecular Formula | C33 H31 Cl F3 N O3 . HCl |
| Formula Weight | 618.52 |
| Physical Form | Solid |
| Product Type | Reagent |
| Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction