missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec 4000 AVANT AIRline

Product Code. 17100775
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Includes:
Attached Socks and Gloves
Socks and Elasticated Boot Overlap
Socks and Neoprene Gloves
Style:
G40T755
G40T755B
G40T755C
G40T755D
Unit Size:
Pack of 3
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Includes Style unitSize
17100775 3X-Large Socks and Elasticated Boot Overlap G40T755D Pack of 3
17120755 Small Socks and Neoprene Gloves G40T755C Pack of 3
17130755 Small Socks and Elasticated Boot Overlap G40T755D Pack of 3
17140755 Medium Attached Socks and Gloves G40T755B Pack of 3
17150755 Medium Socks and Neoprene Gloves G40T755C Pack of 3
17160755 Medium Socks and Elasticated Boot Overlap G40T755D Pack of 3
17170755 Large Attached Socks and Gloves G40T755B Pack of 3
17100755 Small Socks and Elasticated Boot Overlap G40T755 Pack of 3
17120765 X-Large Socks and Elasticated Boot Overlap G40T755D Pack of 3
17180755 Large Socks and Neoprene Gloves G40T755C Pack of 3
17190755 Large Socks and Elasticated Boot Overlap G40T755D Pack of 3
17140765 2X-Large Attached Socks and Gloves G40T755B Pack of 3
17150765 2X-Large Socks and Neoprene Gloves G40T755C Pack of 3
17160765 2X-Large Socks and Elasticated Boot Overlap G40T755D Pack of 3
17130765 2X-Large Socks and Elasticated Boot Overlap G40T755 Pack of 3
17170765 3X-Large Socks and Elasticated Boot Overlap G40T755 Pack of 3
17180765 3X-Large Attached Socks and Gloves G40T755B Pack of 3
17190765 3X-Large Socks and Neoprene Gloves G40T755C Pack of 3
17110755 Small Attached Socks and Gloves G40T755B Pack of 3
17100765 X-Large Attached Socks and Gloves G40T755B Pack of 3
17110765 X-Large Socks and Neoprene Gloves G40T755C Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
This item is not returnable. View return policy
Product Code. 17100775 Supplier Ansell Supplier No. GR40T0075507G05
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Encapsulated chemical protective suits for use with air-fed/supplied systems.

  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress.
TRUSTED_SUSTAINABILITY

Specifications

Color Green
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size 3X-Large
Includes Socks and Elasticated Boot Overlap
No. of Pockets None
Product Type Encapsulated chemical protective suits for use with air-fed/supplied systems
Seam Type Welded and Taped
Style G40T755D
Weight (Metric) 103 gsm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.