Learn More
Ansell™ AlphaTec™ 4000 AVANT AIRline Ultrasonically Welded and Taped G02
Product Code. 17190735
Change view
Click to view available options
Size:
2X-Large
3X-Large
Large
Large/X-Large
Medium
Small
Unit Size:
Pack of 3
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17190735 Large Pack of 3
17130735 Small Pack of 3
17160735 Medium Pack of 3
17120745 Large/X-Large Pack of 3
17150745 2X-Large Pack of 3
17180745 3X-Large Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17190735 Leverantör Ansell™ Leverantörsnummer GR40T0075404G02
Estimated Shipment: 24-06-2027
Lägg i varukorgen
Lägg i varukorgen
missing translation for 'validMainframeMsg'
Denna artikel kan inte returneras. Se returpolicy
AlphaTec™ 4000 AVANT AIRline Ultrasonically welded and taped G02 are affixed by the wearer and detachable for reuse when appropriate. Air flow is adjustable from 340 L/min to 590 L/min at a working pressure of 3.5 to 5 bar.
Affixed by the wearer and detachable.
  • Air flow is adjustable from 340 L/min to 590 L/min at a working pressure of 3.5 to 5 bar.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached sock boots with anti-slip PVC sole - Designed to allow soft or protective safety shoes to be worn inside and offer limited resistance to slip
TRUSTED_SUSTAINABILITY
Specifikationer
Certifications/Compliance CE CATEGORY III (Type 3, EN 1073-1, EN 14594, EN 1149-5)
Color Green
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size Large
Ankle Style Attached Elastic Sock Boots with Anti-slip PVC Sole
Closure Type Zipper
Cuff Style Elasticated double cuffs and permanently attached chemical protective gloves
Disposable Reusable
Gender Unisex
Neck Style Fully encapsulated hood with visor
No. of Pockets None
Protection Class CE CATEGORY III
Resistance Chemical I Particle I Radioactive I ESD
Seam Type Welded and Taped
Sterility Non-sterile
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.