missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 Trousers
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Description
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect
- Silicone free
Specifications
Specifications
| Size | 2X-Large |
| Gender | Unisex |
| Product Type | Chemical-resistant Trousers |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Green |
| Seam Type | Welded and Taped |
| Certifications/Compliance (Europe) | Partial Body Protection Only, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 14126:2003 |
| Style | G40T301 |
| For Use With (Application) | Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction