Learn More
Ansell AlphaTec™ 4000 Trousers
Product Code. p-7228395
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 17140695
Brand: Ansell GR40T0030103
Only null left
Add to basket
Add to basket
A unique multi-layer barrier fabric renowned for its lightweight, yet robust textile feel and exceptional barrier to organic & inorganic chemicals.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect
  • Silicone free
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Gender Unisex
Product Type Chemical-resistant Trousers
Material Multi-layer Barrier Laminate Fabric
Color Green
Seam Type Welded and Taped
Certifications/Compliance (Europe) Partial Body Protection Only, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 14126:2003
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes