Learn More
Ansell™ AlphaTec™ 4000 Ultrasonically Welded and Taped
Product Code. 17180645
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17180645 3X-Large Pack of 10
17130645 Small Pack of 10
17140645 Medium Pack of 10
17150645 Large Pack of 10
17170645 2X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17180645 Supplier Ansell™ Supplier No. GR40T0012107
Add to basket
Add to basket
AlphaTec™ 4000 Ultrasonically welded and taped consists of a unique multi-layer barrier fabric renowned for its lightweight, yet robust textile feel and exceptional barrier to organic and inorganic chemicals.
Consists of a unique multi-layer barrier fabric that provides lightweight and robust textile feel with exceptional barrier to organic and inorganic chemicals.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect, typical innovative features include dual zip systems and double cuffs
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Anti-static, tested according to EN 1149-5
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size 3X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Spruce Green
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Double Inner Knit
Neck Style Hooded
Hood Type 2-panel, Elastic
Closure Type Double Zipper
Seam Type Welded, Taped
Sterility Non-sterile
Protection Class CE CATEGORY III
Resistance Chemicals, Particle
Construction 2-piece Hood, Elasticated Waist, Double Cuffs and Ankles, Double Zip Closure, Double Cuffs with Knitted Inner Cuff, Attached Ansell Barrier Gloves
Collar Type Hood
Certifications/Compliance (Europe) CE CATEGORY III (Type 3-B, Type 4-B, Type 5-B, EN 1073-2, EN 14126, EN 1149-5)
No. of Pockets None
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes