missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 4000 Ultrasonically Welded and Taped Suit
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 2
Pack of 3
Description
- Specialized protection:The AlphaTec™ 4000 Ultrasonically Welded and Taped - Model 130 is a robust chemical protective suit, permeation-tested to withstand more than 200 hazardous chemicals, including chemical warfare agents
- Increased strength: This full body protection suit comes with taped and welded seams to grant users added strength and chemical barrier defense
- Heightened visibility: Designed with orange-colored material, it ensures better visibility which can reduce the risk of workplace injuries
- Improved practicality: It also possesses a chinstrap and a thumb loop, which are compatible with a range of respirators and hand protection products
- Optimized fit: Its elasticated two-piece hood, waist, cuffs and ankles simplify the donning and doffing process
- Certified anti-static capabilities: In addition, this anti-static suit complies with EN 1149-5
- CE Category III Type 3/4/5 protection
Specifications
Specifications
| Color | Orange |
| Hood Type | Attached |
| Material | Multi-layer Barrier Laminate Fabric |
| Size | 4X-Large |
| Ankle Style | Elasticated |
| Closure Type | Single zip closure |
| Collar Type | 3-piece hood |
| Cuff Style | Elasticated |
| Resistance | Chemical |
| Seam Type | Ultrasonicaly welded and taped |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction