Learn More
Ansell AlphaTec™ 4000 Ultrasonically Welded and Taped Suit
Product Code. 18585054 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 2
Pack of 3
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
18585054 3X-Large Pack of 3
18525054 Small Pack of 3
18545054 Medium Pack of 3
18515054 Large Pack of 3
18565054 X-Large Pack of 3
18535054 2X-Large Pack of 3
18555054 4X-Large Pack of 2
18575054 5X-Large Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 18585054 Supplier Ansell Supplier No. OR40T0013007
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AlphaTec™ 4000 Ultrasonically Welded and Taped Suit with innovative multi-layer chemical barrier technology and Type 3/4/5 protection.
  • Specialized protection:The AlphaTec™ 4000 Ultrasonically Welded and Taped - Model 130 is a robust chemical protective suit, permeation-tested to withstand more than 200 hazardous chemicals, including chemical warfare agents
  • Increased strength: This full body protection suit comes with taped and welded seams to grant users added strength and chemical barrier defense
  • Heightened visibility: Designed with orange-colored material, it ensures better visibility which can reduce the risk of workplace injuries
  • Improved practicality: It also possesses a chinstrap and a thumb loop, which are compatible with a range of respirators and hand protection products
  • Optimized fit: Its elasticated two-piece hood, waist, cuffs and ankles simplify the donning and doffing process
  • Certified anti-static capabilities: In addition, this anti-static suit complies with EN 1149-5
  • CE Category III Type 3/4/5 protection
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Hood Type Attached
Material Multi-layer Barrier Laminate Fabric
Size 3X-Large
Ankle Style Elasticated
Closure Type Single zip closure
Collar Type 3-piece hood
Cuff Style Elasticated
Resistance Chemical
Seam Type Ultrasonicaly welded and taped