Learn More
Ansell™ AlphaTec 53-001 Gloves
Product Code. 17353153 Shop All Ansell Products
Change view
Click to view available options
Size: 6; Color: White; Material: Nylon
Product Code.
17353153 Size: 6; Color: White; Material: Nylon
1 options
Product Code. 17353153 Supplier Ansell™ Supplier No. 53001060
  • ONLINE EXCLUSIVE PRICE
    133.30 €
    87.76 € / 6 pairs
    Save 45.54 €
    34% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Chemical protection gloves with nitrile neoprene nitrile chemical gauntlet and Ansell Grip technology
  • These acid-resistant gloves offer a nitrile and neoprene barrier that provides advanced chemical protection from acids to hydrocarbons and beyond
  • These AlphaTec™ 53-001 black chemical resistant gloves have ‘Type A’ chemical protection under EN ISO 374:2016
  • MICROCHEM™ Chemical Barrier Technology combines polymers in proprietary blends or multi-layer structures specific to their application, providing superior protection to wearers
  • ANSELL GRIP™ Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
TRUSTED_SUSTAINABILITY
Specifications
Color White, Black
Latex Free Yes
Material Nylon
Size 6
Style 53-001
Length (Metric) 350 mm