missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Alphatec™ 55-300 Series Neoprene Immersion Gloves

Product Code. 15670914
Change view
Click to view available options
Size: 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15670914 Size: 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15670914 Supplier Ansell Edmont™ Supplier No. 55300090

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides effective protection for medium duty chemical environments. Ansell Edmont™ Alphatec™ 55-300 Series Neoprene Immersion Gloves delivers good chemical resistance against most acids, alcohols, oils, lubricants and hydrocarbons.

  • Chemical and liquid protection gloves
  • Strong physical protection against abrasions and good protection against cuts
  • Medium thickness increases dexterity, which improves the ability to manipulate objects
  • Resistant to Type I allergies
  • Unlined for effective cleaning

Applications:

  • Biotechnologies
  • Blending of liquids
  • Chemical and Medical research
  • Chemical handling
  • Chemical industry
  • Chemicals mixing/handling/transferring/pouring/blending
  • Lab work and analysis, especially when resistance to acids is required
  • Pharmaceuticals
  • Product handling - chemical/biological
  • Product protection applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003,EN 374:2003
Cleanroom Class Non-Critical Environment
Description Grip design: Smooth Finish
Applications Laboratory
Color Black
Latex Free Yes
Material Neoprene
Product Type Neoprene Immersion Gloves
Size 9
Thickness (Metric) 0.51 mm
Glove Cuff Style Gauntlet
Packaging Type 10 pairs in black sealed bags per shipper carton
Port Size (Metric) 150 mm
Series 55-300
Length (Metric) 711 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.