missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 58-008 Extended Full Arm Coverage Gloves

Product Code. 17815641
Change view
Click to view available options
Size:
8
9
10
Unit Size:
10 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17815641 8 10 pairs
17825641 9 10 pairs
17835641 10 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17815641 Supplier Ansell Supplier No. 58008080
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Dry and wet grip enhance dexterity and reduce hand fatigue. Wide cuff to allow donning over protection clothing.

  • Nitrile polymer for excellent mechanical protection as well as resistance to oils, fats, greases and hydrocarbons
  • AlphaTec 58-008 has Type A chemical protection under new ENISO 374:2016
  • Lozenge grip for enhance grip in dry and wet conditions
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 388:2016 +A1:2018, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 388:2016, EN 420:2003 + A1:2009, EN ISO 374-1:2016, EN ISO 374-5:2016, Food Contact
Chlorinated Yes
Accelerator Free No
Applications Industrial
Coating Material Nitrile
Color Green
Latex Free Yes
Material Nitrile
Product Type Protective Gloves
Size 8
Gender Unisex
Glove Cuff Style Extended
Lining Lined
Model 58-008
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.