missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 58-530W Series Nitrile Immersion Gloves

Product Code. 12689741
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
6 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12689741 9 6 pairs
12649741 10 6 pairs
12659741 11 6 pairs
12669741 7 6 pairs
12679741 8 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12689741 Supplier Ansell™ Supplier No. 58535W090
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Recommended for outdoor applications. Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture to prevent possible chemical leakage onto the skin.

  • Nitrile polymer coating reduces chemical leakage onto skin
  • For indoor applications
  • Nylon liner
  • Gauntlet cuff
  • Sea green shell, anthracite gray dip color
TRUSTED_SUSTAINABILITY

Specifications

Back Material Nitrile
Cuff Material Nitrile
Applications Industrial
Coating Material Nitrile
Color Green, Gray
Latex Free Yes
Material Nitrile
Product Type Immersion Gloves
Size 9
Glove Cuff Style Gauntlet
Liner Material Nylon
Lining White Nylon
Quantity 6 Pairs
Resistant To Chemical resistance
Series 58-535W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.