missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 87-085 Lightweight Food-Safe Latex Gloves
Description
- For use in low hazard areas
- Beaded cuff for tear resistance and easy donning
- Resistant, flexible, and comfortable
- 100% cotton flock lining
- AQL 0.65
Specifications
Specifications
| Certifications/Compliance (Europe) | CE 0493, EN 420:2003 + A1:2009, REACH Compliant |
| Color | Pink |
| Hand | Specific |
| Material | Latex |
| Size | 7.5 |
| EN388 Standard | 2010X |
| EN ISO 374-1 | Type B |
| EN ISO 374-1 CHEMICAL LETTER CODES | KLT |
| EN ISO 374 - 5 | Yes |
| Liner Material | 100% Cotton |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?