missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 87-900 Protective Gloves
Description
- Heightened protection: AlphaTec™ 87-900 protective gloves' combination of latex and neoprene ensures protection against a wide range of hazardous chemicals
- Improved durability: Thanks to 0.68 mm/27 mil thickness, these two-toned safety gloves are especially durable and robust, making them suitable for various heavy-duty applications
- Increased comfort: These AlphaTec™ industrial gloves also feature high-quality 100% cotton flock lining, reducing the potential for skin irritation and protecting wearers in comfort
- Enhanced grip: Designed with a honeycomb grip pattern, these heavy-duty work gloves ensure firm, confident grip in both wet and dry conditions
- Assured quality: In addition, they boast an AQL of 1.5
Specifications
Specifications
| Applications | Industrial |
| Color | Green, Yellow |
| Latex Free | No |
| Material | Latex, Neoprene |
| Product Type | Chemical-resistant Gloves |
| Size | 7 |
| Thickness (English) | 16.9 mil |
| Thickness (Metric) | 0.4 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?