missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 88-356 Protective Gloves
Description
- Food handling compliance: Satisfying relevant FDA and EU food safety requirements ( including 21 CFR 177 .2600 ) , AlphaTec™ 87-356 work gloves make for ideal food handling gloves for various food processing applications
- Reduced allergens: They benefit from a special chlorination process, reducing the risk of allergic reactions
- Heightened tactility and sensitivity: With 17 mil/0.43 mm thickness, these AlphaTec™ gloves remain thin, yet durable, with high levels of tactility, sensitivity and dexterity
- Enhanced grip: AlphaTec™ 88-356 light-duty gloves also have a fish scale texture, ensuring excellent grip for both dry and wet handling
- Specialized protection: They also offer EN 374 Type B chemical protection, ideal for low-hazard applications, such as detergent cleaning, as well as mildly acidic, oily food processing
Specifications
Specifications
| Applications | Industrial |
| Color | Blue |
| Finger Thickness (English) | 16.9 mil |
| Finger Thickness (Metric) | 0.43 mm |
| Latex Free | No |
| Material | Rubber Latex |
| Palm Thickness (English) | 16.9 mil |
| Palm Thickness (Metric) | 0.43 mm |
| Product Type | Chemical Protection Gloves |
| Size | 8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?