missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-816 Knitted Nylon Gloves with Fortix™ Nitrile Foam Coating

Product Code. 16099671
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16099671 9 12 pairs
16069671 6 12 pairs
16079671 7 12 pairs
16089671 8 12 pairs
16009681 10 12 pairs
16019681 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16099671 Supplier Ansell Supplier No. 11816090
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

An ultra-thin liner and coating provide a barehand feel for workers who rely on fingertip sensitivity for precision tasks. Fortix™ abrasion resistant foam nitrile coating enhances safety and productivity.

  • Ultra-thin liner and coating ensures a barehand feel for workers who rely on fingertip sensitivity for precise tasks
  • Ergonomic design provides tactility for easy handling of small parts
  • Materials are tested by dermatologists, solvent-free, and Oekotex certified for no harmful substances
  • FORTIX™ Abrasion Resistance Technology extends glove life while enhancing worker safety and productivity
  • No transfer of silicone contaminants to metal parts prior to painting
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Seamless Knit
Coating Material Nitrile
Color Black, Blue
Exterior Finish No Texture
Hand Specific
Latex Free No
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size 9
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, Metal Fabrication, and Transportation
Glove Cuff Style Knit
Series HyFlex™ 11-816
Yarn Gauge 18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.