missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ ESD Gloves 58-001

Product Code. 17874901 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17874901 8 12 pairs
17864901 7 12 pairs
17884901 9 12 pairs
17894901 10 12 pairs
17804911 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17874901 Supplier Ansell Supplier No. 58001080
  • ONLINE EXCLUSIVE PRICE
    132.00 €
    84.73 € / 12 pairs
    Save 47.27 €
    35% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AlphaTec™ 58-001 ESD - The ideal solution to mitigate static buildup when working with chemicals and liquids in environments with static hazards or high explosive risks.

  • AlphaTec™ 58-001 ESD - Unique Nitrile and Carbon formulation consistently dissipates static across the surface and through the glove ensuring consistent passing of EN 1149 and EN 16350
  • High performance
  • Nitrile compound offers a far superior snag, puncture and abrasion protection compared with rubber or Neoprene gloves
  • Reverse lozenge palm finish for improved dry/wet grip
  • Foldable cuff gutter to prevent dripping on the forearm
  • Antistatic flocking layer for ease of donning/doffing and moisture management
  • For Assembly and inspection of components, Oil, fluids and filter change, Plating, coating, sealing, painting, Handling incoming goods, Transferring liquids and solids between vessels and tanks and process equipment, Opening furnaces draining pumps valves or lines and crackers BTX process, Cleaning furnaces, distillation pumps, valves or lines and crackers BTX process
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 388:2016 +A1:2018, EN 420:2003 + A1:2009, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, EN 1149, EN 16350:2014, EN 16350:2014, EN 388:2016, EN ISO 374-1:2016
Accelerator Free No
Applications Industrial
Coating Material Nitrile
Color Green
Cut Resistance Rating ANSI A1
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Laboratory/Cleanroom Use No
Latex Free No
Material Nitrile
Powdered Powder Free
Product Type Glove
Size 8
Sterility Non-sterile
Style Anti-static
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
EN388 Standard 4101X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLOPT
For Use With (Application) Mitigate static buildup
Gender Unisex
Glove Cuff Color Green
Glove Cuff Style Gauntlet
Grip Design Lozenge
Lining Unlined
Model AlphaTec™ 58-001 ESD
Puncture Rating ANSI/ISEA 105-16 Level 1
Series 58-001
Length (English) 13 in.
Length (Metric) 330 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.