Learn More
Ansell™ AlphaTec™MICROCHEM™4000 Model 212 Apron
Product Code. 32177083 Shop All Ansell Products
Change view
Click to view available options
Closure Type:
Waist Ties
Unit Size:
Pack of 20
Product Code. Closure Type Unit Size
32177083 Waist Ties Pack of 20
1 options
Product Code. 32177083 Supplier Ansell™ Supplier No. GR40W0021200
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MICROCHEM™ 4000 is designed to provide anexceptional barrier against organic and inorganicchemicals and biological agents
  • Tie fastening to waist
  • 100cm long tie fastening
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Universal
Material Multi-layer Barrier Laminate Fabric
Color Green
Disposable Single-use
Closure Type Waist Ties
Sterility Non-sterile
No. of Pockets None
Resistance Abrasions, acids, anti-static, bloodborne pathogens, chemicals
Certifications/Compliance EN 14605: 2005 +A1:2009, Type PB (3);EN 14605: 2005 +A1:2009, Type PB (4); EN14126: 2003, Types PB(3)-B, PB (4)-B; EN1149-1: 1995
Length (Metric) 100 cm
Item Description Apron with tie fastenings. Comes in one size only
Style Alphatec 4000, Model 212
Seam Type Welded and Taped
Show More Show Less
Product Title
Select an issue
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.