missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ MICROGARD™ 1500 Coveralls, Model 138
Description
- Three-piece hood
- Elasticated hood, wrists, waist, and ankles
- Two-way front zipper with resealable storm flap
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Small |
| Gender | Unisex |
| Material | SMS Fabric |
| Color | Red |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Neck Style | Hooded |
| Hood Type | 3-panel, Elastic |
| Seam Type | Stitched |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?